VWR® Culture Tubes, Reusable, Borosilicate Glass
Supplier: DWK Life Sciences (KIMBLE)
VW45048-1275
89000-496CS 419.38 USD
89000-496
VWR® Culture Tubes, Reusable, Borosilicate Glass
Tubes Culture Tubes
Tubes are made from Type I, Class A borosilicate glass and feature a durable white ceramic enamel marking spot.
  • Borosilicate glass fabrication
  • Uniform wall thickness
  • Fire-polished top rim
Fire-polished top rim and uniform wall thickness provide maximum heat transfer and chemical resistance.
Certifications: Meet ASTM Specification E982, Type IV requirements.