Water, ARISTAR® PLUS for HPLC low TOC, VWR Chemicals BDH®

Supplier: BDH

10901-VWNA63 87003-652
87003-650CS 988.23 USD
87003-650 87003-652
Water, ARISTAR® PLUS for HPLC low TOC, VWR Chemicals BDH®
Water

For LC, HPLC, and trace organic analysis.


  • Used in analytical and preparative separations
  • Suitable for gradient and isocratic conditions
  • Filtered through a 0.5 μm filter to remove particulates


LC Gradient Suitability Tested:
Column: 25 cm C18
Flow rate: 2 ml/min
Conditions: 40 ml of water are passed through the column and eluted with a water acetonitrile gradient at 5% per min.
Maximum Absorbance: 0.002 at 254 nm

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Color max. 5 APHA
Residue after Evaporation max. 1 ppm
Fluorescence (as Quinine) max. 100 ppt
Resistivity (at time of bottling) min. 18 MΩ
Total Organic Carbon max. 20 ppb
LC Gradient Suitability Test: to pass test
Optical Absorbance at 200 nm max. 0.01
Optical Absorbance at 254 nm max. 0.005
Optical Absorbance at 300 nm max. 0.005
Optical Absorbance at 400 nm max. 0.005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR