VWR® PUREImage™ Cleanroom Paper
Supplier: VWR International
82028-072 82028-074 87003-430 87003-432 87003-434 87003-436 87004-164 87004-170 87004-188 87004-191 87004-192 87004-194 89038-762 94001-262 94001-264 82030-866 89201-138 89201-142 76076-774
82028-072CS 357.84 USD
82028-072 82028-074 87003-430 87003-432 87003-434 87003-436 87004-164 87004-170 87004-188 87004-191 87004-192 87004-194 89038-762 94001-262 94001-264 82030-866 89201-138 89201-142 76076-774
VWR® PUREImage™ Cleanroom Paper
Paper
Cleanroom paper is designed for writing, printing, and photocopying in cleanroom environments where the prevention of chemical exportation and particle contamination is critical. Designed for writing, printing, and photocopying in cleanroom environments.
  • Features high tensile strength and excellent heat resistance for enhanced copier and printing efficiency
  • Web-cleaned on both sides prior to sheeting
  • Polyethylene binding technology results in very low levels of particles, nonvolatile residue (NVR), and ion extractables
  • Unique precision cutting technology
More Product Information