Nitric acid 67 - 70%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Nitric acid solution

10101-VWNQ09 10101-VWCQ09 10101-VWNQ03
87003-261EA 358.86 USD
87003-261 CA87003-261 87003-259
Nitric acid 67 - 70%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Nitric acid

High purity acid for quantitative trace metal analysis at the parts-per-billion (ppb) level.

Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608

Order Now

Specification Test Results

Assay 67 - 70%
Color max. 10 APHA
Aluminum (Al) max. 1 ppb
Antimony (Sb) max. 0.5 ppb
Arsenic (As) max. 0.5 ppb
Barium (Ba) max. 0.1 ppb
Beryllium (Be) max. 0.1 ppb
Bismuth (Bi) max. 0.1 ppb
Boron (B) max. 1 ppb
Cadmium (Cd) max. 0.5 ppb
Calcium (Ca) max. 1 ppb
Cerium (Ce) max. 0.1 ppb
Cesium (Cs) max. 0.1 ppb
Chromium (Cr) max. 1 ppb
Cobalt (Co) max. 0.5 ppb
Copper (Cu) max. 0.5 ppb
Dysprosium (Dy) max. 0.1 ppb
Erbium (Er) max. 0.1 ppb
Europium (Eu) max. 0.1 ppb
Gadolinium (Gd) max. 0.1 ppb
Gallium (Ga) max. 0.1 ppb
Germanium (Ge) max. 0.1 ppb
Gold (Au) max. 0.1 ppb
Hafnium (Hf) max. 0.1 ppb
Holmium (Ho) max. 0.1 ppb
Indium (In) max. 0.1 ppb
Iron (Fe) max. 1 ppb
Lanthanum (La) max. 0.1 ppb
Lead (Pb) max. 0.1 ppb
Lithium (Li) max. 0.1 ppb
Lutetium (Lu) max. 0.1 ppb
Magnesium (Mg) max. 1 ppb
Manganese (Mn) max. 0.1 ppb
Mercury (Hg) max. 0.1 ppb
Molybdenum (Mo) max. 0.1 ppb
Neodymium (Nd) max. 0.1 ppb
Nickel (Ni) max. 0.5 ppb
Niobium (Nb) max. 0.1 ppb
Palladium (Pd) max. 0.5 ppb
Platinum (Pt) max. 0.5 ppb
Potassium (K) max. 1 ppb
Praseodymium (Pr) max. 0.1 ppb
Rhenium (Re) max. 0.1 ppb
Rhodium (Rh) max. 0.5 ppb
Rubidium (Rb) max. 0.1 ppb
Ruthenium (Ru) max. 0.5 ppb
Samarium (Sm) max. 0.1 ppb
Scandium (Sc) max. 0.1 ppb
Selenium (Se) max. 1 ppb
Silver (Ag) max. 0.1 ppb
Sodium (Na) max. 1 ppb
Strontium (Sr) max. 0.1 ppb
Tantalum (Ta) Information Only
Tellurium (Te) max. 0.1 ppb
Terbium (Tb) max. 0.1 ppb
Thallium (Tl) max. 0.1 ppb
Thorium (Th) max. 0.1 ppb
Thulium (Tm) max. 0.1 ppb
Tin (Sn) max. 0.5 ppb
Titanium (Ti) max. 0.5 ppb
Tungsten (W) max. 0.1 ppb
Uranium (U) max. 0.1 ppb
Vanadium (V) max. 0.5 ppb
Ytterbium (Yb) max. 0.1 ppb
Yttrium (Y) max. 0.1 ppb
Zinc (Zn) max. 0.5 ppb
Zirconium (Zr) max. 0.1 ppb
Chloride (Cl-) max. 0.2 ppm
Total Phosphorus (P) max. 0.01 ppm
Total Sulfur (S) max. 0.3 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR