Hydrofluoric acid 47 - 51%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®

Supplier: BDH
Danger

10501-VWNW03
87003-257EA 479.97 USD
87003-257
Hydrofluoric acid 47 - 51%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Hydrofluoric acid

High purity acid for quantitative trace metal analysis at the parts-per-billion (ppb) level.

Formula: HF
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8,II

Order Now

Specification Test Results

Assay 47 - 51 %
Color max. 10 APHA
Aluminum (Al) max. 1 ppb
Antimony (Sb) max. 0.2 ppb
Arsenic (As) max. 0.5 ppb
Barium (Ba) max. 0.1 ppb
Beryllium (Be) max. 0.1 ppb
Bismuth (Bi) max. 0.1 ppb
Boron (B) max. 1 ppb
Cadmium (Cd) max. 0.1 ppb
Calcium (Ca) max. 1 ppb
Cerium (Ce) max. 0.1 ppb
Cesium (Cs) max. 0.1 ppb
Chromium (Cr) max. 1 ppb
Cobalt (Co) max. 0.1 ppb
Copper (Cu) max. 0.5 ppb
Dysprosium (Dy) max. 0.1 ppb
Erbium (Er) max. 0.1 ppb
Europium (Eu) max. 0.1 ppb
Gadolinium (Gd) max. 0.1 ppb
Gallium (Ga) max. 0.1 ppb
Germanium (Ge) max. 0.1 ppb
Gold (Au) max. 0.2 ppb
Hafnium (Hf) max. 0.1 ppb
Holmium (Ho) max. 0.1 ppb
Indium (In) max. 0.1 ppb
Iron (Fe) max. 1 ppb
Lanthanum (La) max. 0.1 ppb
Lead (Pb) max. 0.1 ppb
Lithium (Li) max. 0.1 ppb
Lutetium (Lu) max. 0.1 ppb
Magnesium (Mg) max. 1 ppb
Manganese (Mn) max. 0.1 ppb
Mercury (Hg) max. 1 ppb
Molybdenum (Mo) max. 0.1 ppb
Neodymium (Nd) max. 0.1 ppb
Nickel (Ni) max. 0.5 ppb
Niobium (Nb) max. 0.1 ppb
Palladium (Pd) max. 0.2 ppb
Platinum (Pt) max. 0.2 ppb
Potassium (K) max. 1 ppb
Praseodymium (Pr) max. 0.1 ppb
Rhenium (Re) max. 0.1 ppb
Rhodium (Rh) max. 0.1 ppb
Rubidium (Rb) max. 0.1 ppb
Ruthenium (Ru) max. 0.1 ppb
Samarium (Sm) max. 0.1 ppb
Scandium (Sc) max. 0.1 ppb
Selenium (Se) max. 1 ppb
Silver (Ag) max. 0.5 ppb
Sodium (Na) max. 1 ppb
Strontium (Sr) max. 0.1 ppb
Tantalum (Ta) Information Only
Tellurium (Te) max. 0.1 ppb
Terbium (Tb) max. 0.1 ppb
Thallium (Tl) max. 0.1 ppb
Thorium (Th) max. 0.1 ppb
Thulium (Tm) max. 0.1 ppb
Tin (Sn) max. 0.5 ppb
Titanium (Ti) max. 1 ppb
Tungsten (W) max. 0.5 ppb
Uranium (U) max. 0.1 ppb
Vanadium (V) max. 0.1 ppb
Ytterbium (Yb) max. 0.1 ppb
Yttrium (Y) max. 0.1 ppb
Zinc (Zn) max. 1 ppb
Zirconium (Zr) max. 0.1 ppb
Chloride (Cl-) max. 4 ppm
Fluosilicic Acid (SiF6) max. 20 ppm
Total Phosphorus (P) max. 0.05 ppm
Total Sulfur (S) max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR