Acetic acid ≥99%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®

Supplier: BDH
Danger

10601-VWNQ03 10601-VWNQ09
87003-239EA 228.4 USD
87003-239 87003-241
Acetic acid ≥99%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Acetic acid

High purity acid for quantitative trace metal analysis at the parts-per-billion (ppb) level.

Formula: H₃CCOOH
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay ≥ 99 %
Color max. 10 APHA
Aluminum (Al) max. 1 ppb
Antimony (Sb) max. 0.5 ppb
Arsenic (As) max. 0.5 ppb
Barium (Ba) max. 0.5 ppb
Beryllium (Be) max. 0.1 ppb
Bismuth (Bi) max. 0.1 ppb
Cadmium (Cd) max. 0.5 ppb
Calcium (Ca) max. 1 ppb
Cerium (Ce) max. 0.1 ppb
Cesium (Cs) max. 0.1 ppb
Chromium (Cr) max. 1 ppb
Cobalt (Co) max. 0.1 ppb
Copper (Cu) max. 0.5 ppb
Dysprosium (Dy) max. 0.1 ppb
Erbium (Er) max. 0.1 ppb
Europium (Eu) max. 0.1 ppb
Gadolinium (Gd) max. 0.1 ppb
Gallium (Ga) max. 0.1 ppb
Germanium (Ge) max. 0.5 ppb
Hafnium (Hf) max. 0.1 ppb
Holmium (Ho) max. 0.1 ppb
Indium (In) max. 0.1 ppb
Iron (Fe) max. 1 ppb
Lanthanum (La) max. 0.1 ppb
Lead (Pb) max. 0.1 ppb
Lithium (Li) max. 0.1 ppb
Lutetium (Lu) max. 0.1 ppb
Magnesium (Mg) max. 0.5 ppb
Manganese (Mn) max. 0.5 ppb
Mercury (Hg) max. 1 ppb
Molybdenum (Mo) max. 0.5 ppb
Neodymium (Nd) max. 0.1 ppb
Nickel (Ni) max. 0.5 ppb
Platinum (Pt) max. 0.5 ppb
Potassium (K) max. 1 ppb
Praseodymium (Pr) max. 0.1 ppb
Rhenium (Re) max. 0.1 ppb
Rhodium (Rh) max. 0.5 ppb
Rubidium (Rb) max. 0.1 ppb
Ruthenium (Ru) max. 0.5 ppb
Samarium (Sm) max. 0.1 ppb
Scandium (Sc) max. 0.1 ppb
Selenium (Se) max. 1 ppb
Silver (Ag) max. 1 ppb
Sodium (Na) max. 1 ppb
Strontium (Sr) max. 0.5 ppb
Tellurium (Te) max. 0.5 ppb
Terbium (Tb) max. 0.1 ppb
Thallium (Tl) max. 0.1 ppb
Thorium (Th) max. 0.1 ppb
Thulium (Tm) max. 0.1 ppb
Tin (Sn) max. 0.5 ppb
Titanium (Ti) max. 0.5 ppb
Tungsten (W) max. 0.5 ppb
Uranium (U) max. 0.1 ppb
Vanadium (V) max. 0.5 ppb
Ytterbium (Yb) max. 0.1 ppb
Yttrium (Y) max. 0.1 ppb
Zinc (Zn) max. 1 ppb
Zirconium (Zr) max. 0.1 ppb
Chloride (Cl-) max. 1 ppm
Phosphate (PO₄³-) max. 1 ppm
Substances Reducing Dichromate (K₂Cr₂O₇) to pass test
Substances Reducing Permanganate (KMnO₄) to pass test
Sulfate (SO₄²-) max. 0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR