Water, ARISTAR® ULTRA, ultrapure Trace_metal, HPLC, VWR Chemicals BDH®

Supplier: BDH

Synonyms: Dihydrogen oxide

20901-VWNL63
87003-236CS 1339.05 USD
87003-236
Water, ARISTAR® ULTRA, ultrapure Trace_metal, HPLC, VWR Chemicals BDH®
Water

Ultra high-purity water for quantitative trace metal analysis in the parts-per-trillion (ppt) level.


  • Highly pure water
  • Ideal for metal analysis
  • Quantities measured in parts-per-trillion


Density: 1 g/cm3 (4°C)

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Color max. 10 APHA
Aluminum (Al) max. 20 ppt
Antimony (Sb) max. 10 ppt
Arsenic (As) max. 10 ppt
Barium (Ba) max. 10 ppt
Beryllium (Be) max. 10 ppt
Bismuth (Bi) max. 10 ppt
Boron (B) max. 20 ppt
Cadmium (Cd) max. 10 ppt
Calcium (Ca) max. 10 ppt
Cerium (Ce) max. 10 ppt
Cesium (Cs) max. 10 ppt
Chromium (Cr) max. 10 ppt
Cobalt (Co) max. 10 ppt
Copper (Cu) max. 10 ppt
Dysprosium (Dy) max. 1 ppt
Erbium (Er) max. 1 ppt
Europium (Eu) max. 1 ppt
Gadolinium (Gd) max. 1 ppt
Gallium (Ga) max. 10 ppt
Germanium (Ge) max. 10 ppt
Gold (Au) max. 10 ppt
Hafnium (Hf) max. 1 ppt
Holmium (Ho) max. 1 ppt
Indium (In) max. 1 ppt
Iron (Fe) max. 10 ppt
Lanthanum (La) max. 1 ppt
Lead (Pb) max. 10 ppt
Lithium (Li) max. 10 ppt
Lutetium (Lu) max. 1 ppt
Magnesium (Mg) max. 10 ppt
Manganese (Mn) max. 10 ppt
Mercury (Hg) max. 20 ppt
Molybdenum (Mo) max. 10 ppt
Neodymium (Nd) max. 1 ppt
Nickel (Ni) max. 10 ppt
Niobium (Nb) max. 10 ppt
Palladium (Pd) max. 10 ppt
Platinum (Pt) max. 10 ppt
Potassium (K) max. 10 ppt
Praseodymium (Pr) max. 10 ppt
Rhenium (Re) max. 10 ppt
Rhodium (Rh) max. 10 ppt
Rubidium (Rb) max. 10 ppt
Ruthenium (Ru) max. 10 ppt
Samarium (Sm) max. 10 ppt
Scandium (Sc) max. 10 ppt
Selenium (Se) max. 50 ppt
Silver (Ag) max. 10 ppt
Sodium (Na) max. 10 ppt
Strontium (Sr) max. 10 ppt
Tantalum (Ta) max. 10 ppt
Tellurium (Te) max. 1 ppt
Terbium (Tb) max. 10 ppt
Thallium (Tl) max. 10 ppt
Thorium (Th) max. 1 ppt
Thulium (Tm) max. 10 ppt
Tin (Sn) max. 10 ppt
Titanium (Ti) max. 10 ppt
Tungsten (W) max. 10 ppt
Uranium (U) max. 1 ppt
Vanadium (V) max. 10 ppt
Ytterbium (Yb) max. 10 ppt
Yttrium (Y) max. 1 ppt
Zinc (Zn) max. 10 ppt
Zirconium (Zr) max. 10 ppt
Chloride (Cl-) max. 1 ppb
Phosphate (PO43-) max. 1 ppb
Sulfate (SO42-) max. 1 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR