Nitric acid 67 - 70%, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®

Supplier: VWR International
Danger

87003-658 87003-226 87003-228
87003-658EA 2507.05 USD
87003-658 87003-226 87003-228
Nitric acid 67 - 70%, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®
Nitric acid

Ultra high purity acid for quantitative trace metal analysis at the parts-per-trillion (ppt) level.

Formula: HNO₃
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608

Order Now

Specification Test Results

Assay 67 - 70%
Aluminum (Al) max. 20 ppt
Antimony (Sb) max. 10 ppt
Arsenic (As) max. 20 ppt
Barium (Ba) max. 10 ppt
Beryllium (Be) max. 10 ppt
Bismuth (Bi) max. 10 ppt
Boron (B) max. 10 ppt
Cadmium (Cd) max. 10 ppt
Calcium (Ca) max. 10 ppt
Cerium (Ce) max. 10 ppt
Cesium (Cs) max. 10 ppt
Chromium (Cr) max. 10 ppt
Cobalt (Co) max. 10 ppt
Copper (Cu) max. 10 ppt
Dysprosium (Dy) max. 1 ppt
Erbium (Er) max. 1 ppt
Europium (Eu) max. 1 ppt
Gadolinium (Gd) max. 1 ppt
Gallium (Ga) max. 10 ppt
Germanium (Ge) max. 10 ppt
Gold (Au) max. 20 ppt
Hafnium (Hf) max. 10 ppt
Holmium (Ho) max. 1 ppt
Indium (In) max. 1 ppt
Iron (Fe) max. 10 ppt
Lanthanum (La) max. 1 ppt
Lead (Pb) max. 10 ppt
Lithium (Li) max. 10 ppt
Lutetium (Lu) max. 1 ppt
Magnesium (Mg) max. 10 ppt
Manganese (Mn) max. 10 ppt
Mercury (Hg) max. 50 ppt
Molybdenum (Mo) max. 10 ppt
Neodymium (Nd) max. 1 ppt
Nickel (Ni) max. 20 ppt
Niobium (Nb) max. 1 ppt
Palladium (Pd) max. 20 ppt
Platinum (Pt) max. 20 ppt
Potassium (K) max. 10 ppt
Praseodymium (Pr) max. 1 ppt
Rhenium (Re) max. 10 ppt
Rhodium (Rh) max. 10 ppt
Rubidium (Rb) max. 10 ppt
Ruthenium (Ru) max. 20 ppt
Samarium (Sm) max. 1 ppt
Scandium (Sc) max. 10 ppt
Selenium (Se) Information only
Silver (Ag) max. 10 ppt
Sodium (Na) max. 10 ppt
Strontium (Sr) max. 10 ppt
Tantalum (Ta) Information only
Tellurium (Te) max. 1 ppt
Terbium (Tb) max. 1 ppt
Thallium (Tl) max. 10 ppt
Thorium (Th) max. 1 ppt
Thulium (Tm) max. 1 ppt
Tin (Sn) max. 20 ppt
Titanium (Ti) max. 20 ppt
Tungsten (W) max. 10 ppt
Uranium (U) max. 1 ppt
Vanadium (V) max. 10 ppt
Ytterbium (Yb) max. 1 ppt
Yttrium (Y) max. 1 ppt
Zinc (Zn) max. 10 ppt
Zirconium (Zr) max. 10 ppt


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR