Ammonia solution 20 - 22%, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®

Supplier: BDH
Danger

20701-VWNJ03
87003-214EA 899.02 USD
87003-214
Ammonia solution 20 - 22%, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®
Ammonia solution

Ultra high purity solution for quantitative trace metal analysis at the parts-per-trillion (ppt) level.

Formula: NH₃
Density: 0.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Assay 20 - 22%
Aluminum (Al) max. 20 ppt
Antimony (Sb) max. 10 ppt
Arsenic (As) max. 10 ppt
Barium (Ba) max. 10 ppt
Beryllium (Be) max. 10 ppt
Bismuth (Bi) max. 10 ppt
Cadmium (Cd) max. 10 ppt
Calcium (Ca) max. 10 ppt
Cerium (Ce) max. 10 ppt
Cesium (Cs) max. 10 ppt
Chromium (Cr) max. 10 ppt
Cobalt (Co) max. 10 ppt
Copper (Cu) max. 10 ppt
Dysprosium (Dy) max. 10 ppt
Erbium (Er) max. 10 ppt
Europium (Eu) max. 10 ppt
Gadolinium (Gd) max. 10 ppt
Gallium (Ga) max. 10 ppt
Germanium (Ge) max. 10 ppt
Gold (Au) max. 10 ppt
Hafnium (Hf) Information Only
Holmium (Ho) max. 10 ppt
Indium (In) max. 10 ppt
Iron (Fe) max. 10 ppt
Lanthanum (La) max. 10 ppt
Lead (Pb) max. 10 ppt
Lithium (Li) max. 10 ppt
Lutetium (Lu) max. 10 ppt
Magnesium (Mg) max. 10 ppt
Manganese (Mn) max. 10 ppt
Mercury (Hg) max. 200 ppt
Molybdenum (Mo) max. 10 ppt
Neodymium (Nd) max. 10 ppt
Nickel (Ni) max. 10 ppt
Niobium (Nb) max. 10 ppt
Palladium (Pd) Information Only
Platinum (Pt) Information Only
Potassium (K) max. 10 ppt
Praseodymium (Pr) max. 10 ppt
Rhenium (Re) Information Only
Rhodium (Rh) max. 10 ppt
Rubidium (Rb) max. 10 ppt
Ruthenium (Ru) Information Only
Samarium (Sm) max. 10 ppt
Scandium (Sc) max. 10 ppt
Selenium (Se) Information Only
Silver (Ag) max. 10 ppt
Sodium (Na) max. 20 ppt
Strontium (Sr) max. 10 ppt
Tellurium (Te) max. 10 ppt
Terbium (Tb) max. 10 ppt
Thallium (Tl) max. 10 ppt
Thorium (Th) max. 10 ppt
Thulium (Tm) max. 10 ppt
Tin (Sn) max. 10 ppt
Titanium (Ti) max. 10 ppt
Tungsten (W) max. 10 ppt
Uranium (U) max. 10 ppt
Vanadium (V) max. 10 ppt
Ytterbium (Yb) max. 10 ppt
Yttrium (Y) max. 10 ppt
Zinc (Zn) max. 10 ppt
Zirconium (Zr) max. 10 ppt


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR