StatSpin® Express 3 Centrifuge, Iris Sample Processing
Supplier: Beckman Coulter
SSX3
87003-180EA 11268.19 USD
87003-180
StatSpin® Express 3 Centrifuge, Iris Sample Processing
Centrifuges Benchtop Centrifuges
When rapid serum or plasma is needed, such as in coagulation testing, sample prep for stat chemistry, and cardiac profiles in the ER, this centrifuge is ideal.
  • Settings for 2, 3, and 5 minutes
  • Tubes spun in 2-minute setting equal tubes spun in 7 to 10 minute setting in other centrifuges
  • Up to 8 standard glass or plastic blood-collection tubes accepted
  • Accepted tube size: Between 1.5 to 10 ml
  • Maintenance-free, brushless motor
It has settings for 2, 3, and 5 minutes.
The centrifuge accepts up to eight standard glass or plastic blood-collection tubes from 1.5 to 10 ml. The rugged, brushless motor is maintenance free.
The two-minute setting produces serum or plasma equal to tubes spun for 7 to 10 minutes in conventional clinical centrifuges.
Certifications: UL listed. CE marked.
Ordering information: Includes two-year manufacturer's warranty, 8-place rotor, removable bowl liner, universal power supply, 120 V adapter and power cord, and eight adapters each for 5 and 10 ml tubes.