Steam-Dot™ Indicators, Propper Manufacturing
Supplier: Propper
36400100
87001-938CS 80.09 USD
87001-938
Steam-Dot™ Indicators, Propper Manufacturing
Sterilization Indicators
Steam-Dot™ indicators provide a fast positive visual check for processed packages sterilized in steam.
  • Self-adhesive steam process indicator
  • Permanent white to black color change on exposure to steam
  • Decipher between processed and unprocessed packages
Steam-Dot™ Indicators offer a fast positive visual check for processed steam sterilized items. Steam-Dot™ Indicator changes from white to black after exposure to steam sterilization cycle.
Certifications: AAMI ST60 and ISO 11140-1