Hematoxylin solution Gill II for histology, for cytology

Supplier: POLYSCIENCES INC-RESEARCH CHEM

Synonyms: DcH-48, Natural Black I, Hydroxybrasilin

24243-500
87001-922EA 161.21 USD
87001-922
Hematoxylin solution Gill II for histology, for cytology
Hematoxylin staining solutions

Gold Standard Gill's #2 should be used when a stronger or darker nuclear stain is required for cytology samples or for immunohistochemical counterstaining.


  • Ideal for more intense cytological staining


Gold Standard Series Gill's Hematoxylins are available in three strengths. Choose from Gold Standard Series Gill's Hematoxylin #1, #2, or #3 for optimal staining of your Cytology and Histology samples.

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR