Prussian Blue Iron Stain Kit (Reaction for Demonstration of Iron), Polysciences
Supplier: POLYSCIENCES INC-RESEARCH CHEM
24199-1
87001-921KT 400.21 USD
87001-921
Prussian Blue Iron Stain Kit (Reaction for Demonstration of Iron), Polysciences
Stains Histology Stains
Prussian Blue or Perls’ reaction is used to demonstrate ferric iron and ferritin
This is not a true staining technique, but rather a histochemical reaction. The protein is split off by the hydrochloric acid, allowing the potassium ferrocyanide to combine with the ferric iron. This forms the ferric ferrocyanide or Prussian Blue.