Formaldehyde 16% in aqueous solution, Ultrapure methanol-free
Supplier: POLYSCIENCES INC-RESEARCH CHEM
|
Danger
|
Methanol-free. Suitable for both electron and light microscopy, formaldehyde easily penetrates large blocks of tissue and can be used in combination with glutaraldehyde to fix delicate tissues. Source of formaldehyde is paraformaldehyde.
Formula:
CH₂O Boiling Pt: –19.3 °C (1013 hPa) Melting Pt: –92 °C Density: 0.8153 g/cm³ (–20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 EINECS: 200-001-8 UN: 3334 ADR: 9, Merck Index: 13,04259 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...