Aqua-Poly/Mount, Polysciences
Supplier: POLYSCIENCES INC-RESEARCH CHEM
18606-20
87001-902EA 264.91 USD
87001-902
Aqua-Poly/Mount, Polysciences
Stains Microscopy and staining reagents
Water soluble, non-fluorescing mounting medium formulated for mounting sections from aqueous solutions, frozen sections, and special procedures that will not tolerate alcohols or solvents
Ideal for immunofluorescent techniques as it enhances and retains fluorescent stains. Coverslip directly from water or buffer using standard methods