Protein A Fluoresbrite® YG Microspheres, Polysciences
Supplier: POLYSCIENCES INC-RESEARCH CHEM
17845-1
87001-896EA 1452.69 USD
87001-896
Protein A Fluoresbrite® YG Microspheres, Polysciences
Controls and Calibration Verification Materials
Protein conjugated microspheres are offered as: antibodies, Protein A, and Protein G covalently coupled to fluorescent YG, non-fluorescent, and blue dyed polystyrene microspheres
Protein conjugated microspheres have a nominal diameter of 1.0µm and are packaged as 1.25% solids (w/v) aqueous suspensions. Proteins of user choice can be coupled to microspheres on a custom basis.
Antibody conjugated microspheres can be used to detect trace amounts of specific antigens in solution and to deplete them from solution. Microspheres coupled with Protein A and Protein G will bind to the Fc portion of antibodies raised in most mammals.