Formaldehyde 10% in aqueous solution, Ultrapure methanol-free

Supplier: POLYSCIENCES INC-RESEARCH CHEM
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

04018-1
87001-890EA 494.29 USD
87001-890
Formaldehyde 10% in aqueous solution, Ultrapure methanol-free
Formaldehyde

Methanol-free. Suitable for both electron and light microscopy, formaldehyde easily penetrates large blocks of tissue and can be used in combination with glutaraldehyde to fix delicate tissues. Source of formaldehyde is paraformaldehyde.

Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: –19.3 °C (1013 hPa)
Melting Pt: –92 °C
Density: 0.8153 g/cm³ (–20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 3334
ADR: 9,
Merck Index: 13,04259

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR