Specialized Clamps and Accessories, Thermo Scientific
Supplier: Epredia
715030
16002-302EA 296.08 USD
16002-302 84000-482
Specialized Clamps and Accessories, Thermo Scientific
Clamps General Purpose Clamps
Ensure accuracy and efficiency with Thermo Scientific™ Specialized Clamps and Accessories.
  • For clamping thin materials
  • Spring-loaded jaws secure specimen
  • Compensation slot ensures correct specimen clamping
  • Orienting on the Z-axis
  • Foil clamp is fitted directly on the specimen cylinder
  • Sandwich supporting material is for foil clamp to section thin foil specimens
  • 50 squares of hard material and two strips