Epredia™ HM 450 Sliding Microtome, Thermo Scientific
Supplier: Epredia
910020
84000-418EA 25856.7 USD
84000-418
Epredia™ HM 450 Sliding Microtome, Thermo Scientific
Microtomes and Microtome Assemblies Microtomes
Section paraffin and frozen sections in biological, botanical and materials science applications with the HM 450 Sliding Microtome.
  • Specimen fine orientation on X/Y-axes
  • Knife carrier glides along a maintenance-free precision cross-roller bearing system
  • Memory function for time-saving specimen advance to the knife
  • Ergonomic handle for moving knife carrier
  • Retraction (may be turned off)
  • Three different speed settings for motorized coarse feed
  • Automatic or manual sectioning
This microtome includes automatic fine sectioning and trimming features with selectable specimen retraction
Stepping motor technology assures precise, accurate specimen advancement. Trimming function allows for precision block trimming prior to the sectioning function. User-friendly control panel allows for programming of settings and sectioning parameters. Important sectioning information including section counter, step sum, remaining travel and specimen position memory function are retained.
The integrated knife carrier design of the HM 450 Sliding Microtome ensures fast, comfortable and nontiring operation.
Ordering information: Includes: Integrated removable waste tray, knife holder with integrated, movable knife guard, dust cover, and an instruction manual.
Requires: Standard specimen clamp (715480), available separately.