Fix-Rite 2 Fixative, Epredia™
Supplier: Epredia
76150
84000-142CS 333.74 USD
84000-142
Fix-Rite 2 Fixative, Epredia™
Stains Microscopy and staining reagents
This spray fixative creates a protective coating over cells in routine cytological preparations, allowing slides to be mailed or carried to the laboratory without damaging samples
Packaged in a pocket-sized, non-aerosol spray pump bottle