Paraffin wax blended with synthetic polymers, Solidification point: 55-57°C, Epredia™ Signature Series™ Type H high polymer
Supplier: Epredia
8338
84000-060CS 327.48 USD
84000-060
Paraffin wax blended with synthetic polymers, Solidification point: 55-57°C, Epredia™ Signature Series™ Type H high polymer
Paraffin wax blended with synthetic polymers
Minimize compression and obtain rapid infiltration with Epredia™ Signature Series™ Paraffin, a unique blend of polymers and highly refined paraffin with narrow carbon chain distribution.
  • Triple-filtered to eliminate impurities
  • Type H allows for ultra-thin sections
  • Type L may reduce infiltration times during processing
  • Both Type L and Type H can be used for infiltration and embedding, and have a melting point of 55° to 57°C
Unique blend of polymers and highly-refined paraffin with narrow carbon chain distribution that minimizes compression and offers rapid infiltration. Both Type H and Type L can be used for infiltration and embedding and have a melting point of 55 to 57°C (131 to 135°F). Type H allows for extremely thin sections, while Type L may reduce infiltration times during processing. Both Type H and Type L paraffins are triple-filtered to eliminate impurities and are packaged in easy-to-open, resealable 0.9kg (2lb.) bags
CAS Number: 8002-74-2