Formalin concentrate 4X (10% final concentration) neutral buffered, Epredia™

Supplier: Epredia
Danger

Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol

5700
84000-020CS 322.48 USD
84000-020
Formalin concentrate 4X (10% final concentration) neutral buffered, Epredia™
Formalin

  • Concentrate formula reduces storage space requirement
  • Phosphate buffers are used to stabilize the pH between 6.8 to 7.2
  • Stringent quality control ensures a formaldehyde concentration of 10%
  • Stable to freezing temperatures


Contains neutral phosphate buffer and methanol.


One gallon mixed with 4 gallons of water reconstitutes to 5 gallons of 10% Formalin.

Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR