SmartWall Plus™ Grids and Shelving Accessories, Metro™
Supplier: Metro
WG2448K3 WG3048K3 GS1436K3 FGS1224K3 WG1848K3 GS1836K3 BH6K3 WG1860K3 WG2436K3 GS1448K3 GS1848K3 PGHK6K3 WG1836K3 WG3036K3 GS1830K3
82023-616EA 253.17 USD
82023-616 82023-620 82023-622 82023-632 83009-612 82023-628 82023-636 83009-614 82023-614 82023-624 82023-630 82023-634 83009-610 82023-618 82023-626
SmartWall Plus™ Grids and Shelving Accessories, Metro™
Graticules, Reticles and Grids
Grids and shelving accessories allow users to organize SmartWall Plus storage systems to precise specifications.
  • Grids maintain a total unit weight capacity of 114 kg (250lbs.)
Storage options can be easily repositioned along the track before securing them to the wall to maximize storage space and improve organization. Metroseal three finishes provide Microban® built-in product protection.
Ordering information: Accessory bracket kits for wire grids (82023-344) include two top brackets that attach to track and four bottom brackets that attach to wall surface. One kit is required per grid. Brackets are constructed of Type 304 stainless steel. Bracket kits for Starsys® wall cabinets (82023-334) include two top brackets and one bottom bracket. Top brackets are Type 304 stainless steel and bottom brackets are steel coated with corrosion-resistant taupe epoxy.