Super Erecta® Wire Shelving, Metroseal 3™ Finish, Metro™
Supplier: Metro
1424NK3 1448NK3 1824NK3 1836NK3 1848NK3 1860NK3 2124NK3 2148NK3 2460NK3 1460NK3 1830NK3 1872NK3 2136NK3 2142NK3 2172NK3 2424NK3 2448NK3 3048NK3 1430NK3 1436NK3 1442NK3 1472NK3 1842NK3 2154NK3 2430NK3 2436NK3 2442NK3 2454NK3 3060NK3 3072NK3 3660NK3 3672NK3 1854NK3 2130NK3 2472NK3 3036NK3 3636NK3 3648NK3 2160NK3
83009-332EA 118.58 USD
83009-332 83009-340 83009-346 83009-350 83009-354 83009-358 83009-362 83009-370 83009-390 83009-342 83009-348 83009-360 83009-366 83009-368 83009-376 83009-378 83009-386 83009-396 83009-334 83009-336 83009-338 83009-344 83009-352 83009-372 83009-380 83009-382 83009-384 83009-388 83009-398 83009-400 83009-410 83009-412 83009-356 83009-364 83009-392 83009-394 83009-406 83009-408 83009-374
Super Erecta® Wire Shelving, Metroseal 3™ Finish, Metro™
Furniture Shelves
Design your own wire shelf system with appropriate accessories.
  • Metroseal 3™ shelves are an economical alternative to stainless steel
  • Corrosion-resistant shelves are electroplated carbon steel, with a special epoxy coating
Open-wire design minimizes dust accumulation, allows a free circulation of air, and provides greater visibility of stored items. Easy shelf adjustment is possible through the use of SiteSelect® stationary or mobile posts (sold separately), which allow shelves to be readjusted and snapped into place at precise 2.5 cm (1") increments along the entire height of the post. An easily identifiable double groove is positioned every 20.3 cm (8") to further aid shelf alignment. Stationary posts are fitted with adjustable leveling bolts for use on uneven surfaces. Mobile posts require the purchase of stem casters. Stem casters (see 19766-236 series) and shelf dividers and ledges (see 22233-212 series) are available separately.
Shelves up to 121.9 cm (48") in length can support up to 363 kg (800 lbs.) per level. Shelves longer than 121.9 cm (48") and all super wide shelves can support up to 272 kg (600 lbs.) per level.
Note: The 35.6 cm (14") wide shelves require the use of footplates (see 19766-814 series) when used as a freestanding unit. The actual length of the shelves is 3.2 mm (¹/₈") shorter than the nominal dimension shown. Stationary posts over 189.6cm high and mobile posts over 138.6 cm high are not for use with 35.6 cm wide shelves. The height of the chrome-plated and stainless steel stationary posts includes a leveling bolt and cap.
Metroseal 3™ shelves includes antimicrobial product protection, which inhibits the growth of bacteria, mold, mildew, and fungi that cause odors, stains, and product degradation. Use within coolers to resist corrosion from moisture. Shelves are not cart-washable.
Certifications: Shelves meet MIL-S-40144E specifications.
Ordering information: Each shelf and post is sold separately. To order posts or accessories, contact your Avantor representative for more information. The exclusive plating-plus-coating system has a 12-year limited warranty against rust formation. Each shelf includes four pairs of plastic split sleeves for securing shelf to posts.