HBIO 96-Well Disposable Electroporation Plates, Coated, BTX
Supplier: Harvard Apparatus
45-0450
83009-294EA 487.93 USD
83009-294
HBIO 96-Well Disposable Electroporation Plates, Coated, BTX
Electroporation Plates
BTX High throughput electroporation plates are sterile multi-well electroporation plates for transfection/transformation of bacteria, yeast, insect, plant and mammalian cells.
  • 2 mm gap
  • 100 µl
  • For use with ECM 630 and ECM 830 Electroporation systems
Try out many different electroporation conditions (cell density, buffer, pulse voltage and width) to find the highest transfection efficiency, quickly and easily, with the ECM 830 HT electroporation system, the ECM 630 HT electroporation system, or the Gemini X2 electroporation system. The disposable, sterile multi-well plate is the heart of the system. It’s like 96 electroporation cuvettes in one convenient plate.