Reagent Bottles with Stoppers, Polypropylene, Wide Mouth, BrandTech
Supplier: Brandtech
101794
83008-546PK 206.19 USD
83008-546
Reagent Bottles with Stoppers, Polypropylene, Wide Mouth, BrandTech
Bottles Roller Bottles
Polypropylene reagent bottles with polypropylene stoppers provide excellent chemical resistance to many aliphatic alcohols, aldehydes, aliphatic hydrocarbons, alkalis, and acids.
  • Break-resistant
  • Polypropylene bottle and stopper for high chemical resistance
  • High translucency
  • Rugged design
Bottles and stoppers are autoclavable.