OPTI-LYNX™ Holder Tube for Trap/Filter Inserts
Supplier: Optimize
11-02570-AA
83008-174EA 100.19 USD
83008-174
OPTI-LYNX™ Holder Tube for Trap/Filter Inserts
Chromatography Supplies Chromatography Spare Parts
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances.
  • For use with OPTI-LYNX™ hardware
  • Installs into holder cap to make connections with trap/filter cartridges
  • Modifies an OPTI-LYNX™ holder to accept trap/filter cartridges
Installs into holder cap to make connections with OPTI-LYNX™ trap cartridges and filter inserts.