Eyewash System Combination Eyewash/Sink Faucet, Speakman®
Supplier: Speakman
SEF-9000
83008-084EA 1902.75 USD
83008-084
Eyewash System Combination Eyewash/Sink Faucet, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
System combines an eyewash/drench hose with a service sink faucet.
This creative design allows the eyewash to be attached directly to the faucet’s cold water integral stop cavity and permits both units to be operated independently. As a result, only one plumbing connection is required and there is never a worry about delivering scalding water to the eyes.
Certifications: Meets ANSI Z358.1 standard.