Pipetus® Pipette Filler, Hirschmann
Supplier: Hirschmann
9907200
83007-688EA 775.44 USD
83007-688
Pipetus® Pipette Filler, Hirschmann
Pipettes Pipette Controllers
Pipetus® Pipette filler are battery powered and suitable for samples ranging from 0.1 to 200 ml.
  • Pump motor emits low noise levels and is free of vibration
  • Highly accurate pipetting
  • Pipette seat is sealed absolutely tight with a silicone adapter
  • Hydrophobic PTFE diaphragm filter with 0.2 µm pore size
  • NiMH batteries
Charging and dispensing of the medium is realised at the press of a button. Exact meniscus configuration is enabled through fine dispensing valves with valve pins made of ceramic compound. The valves are free of wear and offer maximum chemical resistance. An exact and continuously adjustable pumping and suctioning speed can be selected for both filling and venting within five precisely harmonised performance stages. The integrated color display shows the pipetting mode, speed levels, and charging status of the battery. Battery charging is realised without contacts and controlled by a microprocessor in an inductive charging station. The sophisticated ergonomic shape aids non-fatiguing work.