Stainless Steel Undermount Sink, Just Manufacturing
Supplier: Just Manufacturing
US1821AGR
83007-406EA 859.37 USD
83007-406
Stainless Steel Undermount Sink, Just Manufacturing
Furniture Casework
This undermount sink features seamless die-drawn construction of 18-gauge, Type 304 stainless steel.
  • Interior surfaces are polished to a nonporous hand-blended finish
  • Fully-coated underside insulates for sound and reduces condensation
  • Straight-sided compartment provides greater capacity
Certifications: Conforms to ASME/ANSI A112.19.3M.