TAE Buffer, 10X, Molecular Biology Grade, Calbiochem®, MilliporeSigma
Supplier: MilliporeSigma
574797-1L
82604-016EA 173.3 USD
82604-016
TAE Buffer, 10X, Molecular Biology Grade, Calbiochem®, MilliporeSigma
Buffers Electrophoresis Buffers Running Buffers
10X concentrate that can be diluted to a 1X solution containing 40 mM Tris, 40 mM acetate, and 1 mM EDTA, pH ~8.3.