GANT58 ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 4,4',4'',4'''-(Thiophene-2,3,4,5-tetrayl)tetrapyridine, 4-(2,4,5-Tri(pyridin-4-yl)thiophen-3-yl)pyridine, NSC 75503, Hh/Gli Antagonist, GANT 58

M-Clarity™ quality level = MQ100

373400-5MG
82603-616EA 301.28 USD
82603-616
GANT58 ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
GANT58

The Hh/Gli Antagonist, GANT58, also referenced under CAS 64048-12-0, controls the biological activity of Hh/Gli. This small molecule/inhibitor is primarily used for Cell Signaling applications.


Packaging: Packaged under inert gas

Formula: C₂₄H₁₆N₄S
MW: 392.4756 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01103196
CAS Number: 64048-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR