Tetradecyltrimethylammonium bromide ≥99% (by titration), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: MiTMAB, (1-Tetradecyl)trimethylammonium bromide, N,N,N-Trimethyltetradecan-1-aminium bromide, Tetradonium bromide, Tetradecyl trimethyl ammonium Bromide, Myristyltrimethylammonium bromide, Dynamin Inhibitor II, TTAB

M-Clarity™ quality level = MQ100

324411-500MG
82603-536EA 87.43 USD
82603-536
Tetradecyltrimethylammonium bromide ≥99% (by titration), Calbiochem®, Sigma-Aldrich®
Tetradecyltrimethylammonium bromide

The Dynamin Inhibitor II, MiTMAB, also referenced under CAS 1119-97-7, controls the biological activity of Dynamin. This small molecule/inhibitor is primarily used for Membrane applications.


A cell-permeable cationic surfactant that reversibly inhibits dynamin GTPase activity (IC50 = 3.15 µM) by targeting the PH domain and interfering with phospholipid binding. Shown to block dynamin I-dependent SVE (synaptic vesicle endocytosis) in rat brain synaptosomes and dynamin II-dependent RME (receptor-mediated endocytosis) in non-neuronal HeLa, A431, and COS-7 cells. Inhibition is competitive with respect to PS and noncompetitive with respect to GTP.

Formula: C₁₇H₃₈BrN
MW: 336.394 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011770
CAS Number: 1119-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR