Polymyxin B sulphate in aqueous solution cell culture tested, sterile-filtered

Supplier: MilliporeSigma

420413-20ML
82603-000EA 427.41 USD
82603-000
Polymyxin B sulphate in aqueous solution cell culture tested, sterile-filtered
Polymyxin B sulphate
Formula: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
MW: 1385.61 g/mol
MDL Number: MFCD00131991
CAS Number: 1405-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR