Insect Breeding Containers, Greiner Bio-One
Supplier: Greiner Bio-One
217101
82050-328CS 126.47 USD
82050-328
Insect Breeding Containers, Greiner Bio-One
Vials
These polystyrene breeding containers are specifically designed for the handling, storage, and transportation of living insect specimens.
  • Hydrophobic
  • Nonsterile
  • Lighter and stronger than glass
The clear polystyrene tubes are lighter and stronger than glass, yet provide the same optical clarity. The hydrophobic nature of polystyrene prevents moisture accumulation, maintaining dry specimen storage.
White ceaprene foam plugs (sold separately) permit unrestricted gas exchange.