Immuno Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
115071
82050-126CS 235.32 USD
82050-126
Immuno Tubes, Greiner Bio-One
Tubes Immunology Tubes
Immuno tubes are designed for all solid phase assay applications, including TSH (thyroid stimulating hormone) or other hormone determinations.
  • Available in a range of binding capacities
  • Polyethylene or polystyrene tube construction
  • Low-binding tubes are biologically inert and have excellent chemical resistance
  • Excellent optical clarity in medium- and high-binding models
Tubes are available in round- or star-bottom formats. Star-bottom tube interiors increase the available surface area to enhance levels of antigen or antibody binding. Designed for compatibility with standardized equipment, tubes are available in 4 or 5 ml volumes with a selection of low-, medium-, or high-binding capacities. Low-binding polyethylene tubes are biologically inert and offer excellent chemical resistance. Medium- and high-binding polystyrene tubes offer excellent optical clarity.