Neoprene Glove Box Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
8N3032A/10H 5N3032/9Q 8N3032/8H 8N3032AK/9Q 8N1532/8H 8NLL3032/10H 7NLL3032/9Q 8NLL3032/9Q
56223-145CS 9012.55 USD
56223-145 82031-220 82031-314 56223-147 56223-149 56223-150 56223-206 56223-210 82031-216 82031-246 82031-312 56223-208 56223-212 82031-196 82031-226 82031-244 82031-248 82031-270 82031-310 94002-630 82031-262 82031-268 56223-143
Neoprene Glove Box Gloves, Honeywell Safety
Gloves
Premium quality milled neoprene gloves offer high tensile strength and flexibility.
  • Seamless, one-piece construction provides maximum strength and integrity
  • Ideal for applications requiring good sensitivity and dexterity
  • Lead-loaded gloves contain 0.1 mm lead equivalent for attenuation of soft gamma radiation
Ordering information: Butyl, Hypalon®, Neoprene/Hypalon® blend, and Viton® glove box gloves are also available (see 82031-206 series).