Jordan Ultra-Wide Mini-Submarine System, IBI Scientific
Supplier: IBI Scientific
JSB-96
82031-126EA 1052.52 USD
82031-126
Jordan Ultra-Wide Mini-Submarine System, IBI Scientific
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
This electrophoresis system allows quick-screening of large quantities of samples with limited agarose usage
The system is ideal for PCR applications, RFLP analysis, and plasmid screening. Constructed of durable, three-piece cast acrylic. Microtiter plate format can run samples, standards, and blanks in under 90 minutes. Comb 82031-142 is compatible with most multichannel pipettors.
Ordering information: System includes UV transmittant casting tray, casting fixture, one 1.5mm 25-tooth comb, one 1.5mm 50-tooth comb, comb holder, bubble level, adjustable feet, power cords, and manual.