Nunc® Polyester Acrylic Sealing Tape and Applicator, Thermo Scientific
Supplier: Thermo Fisher Scientific
236707
82030-936CS 1224.48 USD
82030-936
Nunc® Polyester Acrylic Sealing Tape and Applicator, Thermo Scientific
Seals Microplate Seals
The clear polyester acrylic tape is pierceable by pipette.
The tape applicator dispenses a polyester tape with acrylic adhesive quickly and efficiently for all general-purpose lab applications. The applicator ensures faster application time compared to tape administered in loose sheets, and can be easily stored in the lab. Applicator dimensions: 121×78 mm.
Ordering information: Tape is supplied on a roll of 1000 sheets within the applicator case.