Puritan® Round Polyester Tipped Applicator, Acetal Handle, Puritan Medical Products
Supplier: Puritan Medical Products
3625
82030-712CS 131.64 USD
82030-712 82030-714
Puritan® Round Polyester Tipped Applicator, Acetal Handle, Puritan Medical Products
Swabs Swabs for Sampling
Symmetrical tip is free from volatile residues, particles, contaminating adhesives, and lint
Ideal for removing epoxy residues from connectors and for use as a cleaning swab for fiber optics compontent manufacturing. Acetal shaft. Made in the USA.
Certifications: CE marked.
Ordering information: Swabs are packaged 100 per pack, 10 packs per case.