Puritan® Medium Round Polyester Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
3605
82030-706CS 1085.1 USD
82030-706
Puritan® Medium Round Polyester Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Sampling
For use in critical environment cleaning
Chemical-resistant, nonabrasive, symmetrical tip is free from volatile residues, particles, contaminating adhesives, and lint. Polypropylene shaft. Made in the USA.
Certifications: CE marked.
Ordering information: Swabs are packaged 100 per pack, 10 packs per case.