Nunc™ Screwdriver for Cryobank and Bank-It™ Vial Systems, Thermo Scientific
Supplier: Thermo Fisher Scientific
369643
82030-534EA 84.63 USD
82030-534
Nunc™ Screwdriver for Cryobank and Bank-It™ Vial Systems, Thermo Scientific
Screwdrivers
Quickly and easily cap and decap Nunc Cryobank tubes using the manual Nunc™ Screwdriver for Cryobank and Bank-It™ Systems
Nunc Cryobank vials provide the customer with superior tracking of their cryovials using a 2D code, the highest storage density on the market and the ability to automate the capping and decapping of the vials. The tubes click into the rack and can be handled without the risk of accidentally dropping a vial.