VWR® Microbiology/Urinalysis Specimen Containers
Supplier: VWR International
82030-366
82030-366CS 265.98 USD
82030-366
VWR® Microbiology/Urinalysis Specimen Containers
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
These clear, 100% virgin medical-grade polypropylene containers are specifically designed for the collection and transport of microbiology, urinalysis, and drug testing specimens.
  • Medical-grade polypropylene fabrication
  • Clear siding
  • Screw-thread caps provide a leak-resistant seal
Clear design offers exceptional clarity
Containers are supplied with the SURE-LOK® proof-of-closure system. Screw-thread polyethylene caps provide a leak-resistant seal. Molded-in features on both the cap and container provide visual and tactile assurance of a secure seal. When the indicator on the cap aligns at or beyond the indicator on the container base, the container is firmly closed. Containers are labeled with a security tab.
Note: Containers are intended for single use, although the container base may be autoclaved at 121°C (250°F). Caps are not autoclavable.
Containers are crack-resistant and feature molded-in graduation markings on three sides. The top and bottom of each container is equipped with a positive grip-knurl to ease in opening or closing when handled with gloves. The straight-sided construction increases stability and allows containers to be stacked.
Certifications: Containers are 95kPa approved.