Nunc® 384 Well Polystyrene Immobilizer Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
436017
82029-702CS 913.28 USD
82029-702
Nunc® 384 Well Polystyrene Immobilizer Plates, Thermo Scientific
Microplates Immunology Microplates and Strips
Nunc 384-Well Polystyrene Immobilizer Plates offer amino-coated and streptavidin surfaces
These plates are optimized for scintillation counting, and have flat bottom wells for optical reading. They have rounded-square wells which eliminates wicking.