Puritan® Wound Measuring Device/DM Stick, Puritan Medical Products
Supplier: Puritan Medical Products
25-15061PFDM
82029-066CS 506.13 USD
82029-066
Puritan® Wound Measuring Device/DM Stick, Puritan Medical Products
Swabs Swabs for Clinical Use
Sticks consist of a 15.2cm (6") polystyrene shaft with an indented foam tip
Made in the USA.
The shaft has a graduated measurement scale for accurate wound measurement. Wound measurement stick 82029-066 is sterile.
Certifications: CE marked.