Retic-Chex® Linearity Controls, Streck
Supplier: STRECK LABORATORIES
285535
82030-690PK 604.68 USD
82030-690 82028-952
Retic-Chex® Linearity Controls, Streck
Controls and Calibration Verification Materials
Controls are designed to establish the patient reportable range and the analytic linearity of the system
Retic-Chex stain is buffered and filtered new methylene blue that can be used with control and patient samples. It is packaged in a 3 mL plastic squeeze dropper vial and can be stored at room temperature.
Available in five-vial sets with values assigned. Open-vial stability is five days and closed-vial stability is 75 days. Compatible with Abbott®, Beckman Coulter®, and Sysmex® instruments.
Ordering information: Each control is supplied with five vials, one each for levels 1, 2, 3, 4 and 5. The 3.0 mL control is designed specifically for Beckman Coulter® LH 750/LH 755 instruments.