PICKPAK™ First Aid PICK Kits, PICK® International
Supplier: Pick International
24PWAC2
82028-888CS 315.3 USD
82028-888 82028-890
PICKPAK™ First Aid PICK Kits, PICK® International
First Aid Supplies First Aid Kits
The First Aid PICK™ Kits are available in basic or advanced kit configurations, and include procedure packs and unitized supplies to meet ANSI guidelines.
First aid cases are constructed of durable polyurethane with a rubber-lined seal. Empty cases are also available separately, allowing for custom-built first aid configurations. In order to maintain compliance with treatment certification levels, case exteriors are clearly marked, specifying basic or advanced treatment contents.
Both basic and advanced kits are stocked with components that correspond to the appropriate level of treatment certification taught in common first aid training. Contents are packaged in order of applicable use, and every package includes personal protection, treatment supplies, and biohazard disposal items designed to reduce the threat of infectious disease or harmful bloodborne pathogens spread during treatment. Unitized loose supplies (82028-904) are pre-arranged for easy inventory and restocking to maintain compliance.
Ordering information: For additional information on PICKPAK treatment-specific pack contents, see series 82028-858 (Burn, Optic, Insect Sting, and Wound Pak), 82028-876 (Bio-Quik Spill Pak), or 82028-870 (CPR Pak). Unitized refill (82028-904) contains 1 cold pack, 16 adhesive bandages (1x3"), 1 triangular bandage with safety pins, 1 pair of forceps, 1 pair of scissors, and 4 packets of burn gel.