VWR® Micro Spoon, Stainless Steel
Supplier: VWR International
82027-526
82027-526CS 676.54 USD
82027-526
VWR® Micro Spoon, Stainless Steel
Spoons
Designed for use in the transfer and handling of small chemical or biological samples.
  • Rustproof
  • Stainless steel construction
The rustproof micro spoon is constructed of stainless steel and features a plastic handle. Substances will not stick to spoon surface.