VWR® Waugh Forceps
Supplier: VWR International
82027-428
82027-428CS 3552.49 USD
82027-428
VWR® Waugh Forceps
Forceps
Waugh-type forcep with 1x2 teeth and guide pin.
  • Stainless steel construction
  • Waugh-type design
  • Teeth help ensure a firm grip
Forceps are straight and feature cross-serrated tips.