VWR® Hemostatic Forceps, Serrated Tip
Supplier: VWR International
82027-416
82027-416CS 1676.41 USD
82027-416
VWR® Hemostatic Forceps, Serrated Tip
Forceps
Hemostatic forceps are used in many surgical procedures, usually to control bleeding or the flow of liquids in tubing.
  • Stainless steel fabrication
  • Serrated tips
These forceps have ring handles just like scissors, and ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.